Learn More
Sigma Aldrich Fine Chemicals Biosciences Deoxyribonucleic acid sing
Catalog No. 502902874
Encompass
A ready-to-use SOLUTIONon of high quality single-stranded template DNA isolated from the testes of salmon. The SOLUTIONon is supplied at a concentration of 9 - 12 mg/ml .
Catalog No. 50-290-2874 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D91565X1ML
Add to Cart
Add to Cart
A ready-to-use SOLUTIONon of high quality single-stranded template DNA isolated from the testes of salmon. The SOLUTIONon is supplied at a concentration of 9 - 12 mg/ml .