Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonucleic acid sodium salt, calf thymus

Catalog No. AAJ64400MC
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64400MC 100 mg
AAJ6440003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64400MC Supplier Thermo Scientific Chemicals Supplier No. J64400MC

You must be signed in and have a business account to purchase online.

missing translation for 'validMainframeMsg'

Suitable for use as a substrate in DNase assays | CAS: 91080-16-9

Deoxyribonucleic acid sodium salt, is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxyribonucleic acid sodium salt, is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.

Solubility
Soluble in water.

Notes
Highly polymerized. Hyperchromicity is at least 27%.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91080-16-9
MDL Number MFCD00163436
IUPAC Name Deoxyribonucleic acid sodium salt

Specifications

Color White
Quantity 100 mg
Solubility Information Soluble in water.
Physical Form Lyophilized Powder
Chemical Name or Material Deoxyribonucleic acid sodium salt, calf thymus
EINECSNumber 293-507-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.