Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonucleic Acid Sodium Salt, Fisher BioReagents

Catalog No. p-25437 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP25141 1 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP25141

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Deoxyribonucleic Acid Sodium Salt is used in several biochemical research applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68938-01-2
Molecular Formula C17H15BrFNO3
Molecular Weight (g/mol) 50000-100000
MDL Number MFCD00130922
InChI Key GXZCYECDYOBPGH-UHFFFAOYNA-N
PubChem CID 73904662
IUPAC Name (2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
SMILES *

Specifications

Color White
Loss on Drying 14% max.
Packaging Amber Glass
Quantity 1 g
Formula Weight 6000000
Physical Form Solid
Chemical Name or Material Deoxyribonucleic Acid Sodium Salt, From Salmon Sperm
Safety handling Check Alert storage code Red
Recommended Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.