Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonucleic Acid Sodium Salt from Salmon or Herring Sperm , MP Biomedical

Catalog No. MP215227580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215227580 100 g
MP215227510 10 g
MP215227525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215227580 Supplier MP Biomedicals, Inc Supplier No. 0215227580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deoxyribonucleic acid sodium salt has been used in a study to evaluate the cytotoxicity of enzyme inhibitors and absorption enhancers in Caco-2 cells for oral delivery of salmon calcitonin.

  • Deoxyribonucleic acid sodium salt has been used in a study to develop a simple method for extracting DNA from Cryptosporidium oocysts
  • Deoxyribonucleic Acid is a polynucleotide
  • It is an essential component of chromosomes in cell nuclei
  • It is the carrier of genetic material
  • It contains information in chemical code to direct the development of the cell according to its inheritance
  • The purine and pyrimidine bases of the nucleosides are primarily adenine, guanine, cytosine and thymine
  • The sugar is D-2-deoxyribose
  • The nucleosides are linked together by phosphates in diester linkage from the 3'-hydroxyl of one sugar to the 5′-hydroxyl of the next
  • The repeating sugar-phosphate linkage forms the backbone of the single polynucleotide strand which is the primary structure of DNA
  • Chemical analysis of DNA from different species show that the purine content is equal to the pyrimidine content; adenine content equal to thymine; guanine equal to cytosine
  • DNA also acts as a template in the formation of ribonucleic acids, which play a fundamental role in the synthesis of proteins in the cell
TRUSTED_SUSTAINABILITY

Specifications

CAS 9007-49-2
Color Cream
Packaging Glass
Quantity 100 g
Grade Not Available
Physical Form Powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.