Learn More
Medchemexpress LLC HY-N2187 5mg Medchemexpress, Deoxyshikonin CAS:43043-74-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21875MG
Encompass_Preferred
Medchemexpress, HY-N2187 5mg Deoxyshikonin CAS:43043-74-9 Deoxyshikonin is isolated from Lithospermum erythrorhizon Sieb with antitumor activity. Deoxyshikonin increases the expression of VEGF-C and VEGF-A mRNA in HMVEC-dLy, promotes HIF-1α and HIF-1β subunit interaction and binds to specific DNA sequences targeted by HIF, indicates a prolymphangiogenesis as well as a proangiogenesis effect in vitro. Deoxyshikonin shows significant synergic antimicrobial activity against S. pneumonia (MIC=17 μg/mL), also shows significantly inhibitory activities against MRSA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0374