Learn More
Revvity Health Sciences Inc Deoxythymidine 5'-Triphosphate, [Methyl-3H]-, Tetrasodium Salt, Specific Activity: 70-90 Ci (2.59-3.33 TBq)/mmol, 250 µCi (9.25 MBq)
Catalog No. 502810978
Encompass
Deoxythymidine 5'-Triphosphate, [Methyl-3H]-, Tetrasodium Salt, Specific Activity: 70-90 Ci (2.59-3.33 TBq)/mmol, 250 µCi (9.25 MBq)
Catalog No. 50-281-0978 Supplier Revvity Health Sciences Inc Supplier No. NET221A250UC
Deoxythymidine 5'-Triphosphate, [Methyl-3H]-, Tetrasodium Salt, Specific Activity: 70-90 Ci (2.59-3.33 TBq)/mmol, 250 µCi (9.25 MBq)