Learn More
Revvity Health Sciences Inc Deoxythymidine 5'-Triphosphate, [Methyl-3H]-, Aqueous Sol, Tetrasodium Salt, Specific Activity: 70-90 Ci (2.59-3.33 TBq)/mMole, 1 mCi (37 MBq)
Catalog No. 502810629
Encompass
Deoxythymidine 5'-Triphosphate, [Methyl-3H]-, Aqueous Sol, Tetrasodium Salt, Specific Activity: 70-90 Ci (2.59-3.33 TBq)/mMole, 1 mCi (37 MBq)
Catalog No. 50-281-0629 Supplier Revvity Health Sciences Inc Supplier No. NET221A001MC
Add to Cart
Add to Cart
Deoxythymidine 5'-Triphosphate, [Methyl-3H]-, Aqueous Sol, Tetrasodium Salt, Specific Activity: 70-90 Ci (2.59-3.33 TBq)/mMole, 1 mCi (37 MBq)