Learn More
Cayman Chemical DeoxyvIolaceIn 2.5mg
SDP
Catalog No. 502995131
Encompass_Preferred
A bacterial metabolite with anticancer properties; inhibits proliferation of hepatocellular carcinoma cells when used at concentrations ranging from 0.1 to 1 µM; has antibacterial activity against Gram-positive bacteria when used at a concentration of 125 UG/ml; has antifungal activity when used at a concentration of 2 mg/ml
Catalog No. 50-299-5131 Supplier Cayman Chemical Supplier No. 234772.5 MG
A bacterial metabolite with anticancer properties; inhibits proliferation of hepatocellular carcinoma cells when used at concentrations ranging from 0.1 to 1 µM; has antibacterial activity against Gram-positive bacteria when used at a concentration of 125 UG/ml; has antifungal activity when used at a concentration of 2 mg/ml