Learn More
Selleck Chemical LLC Depatuxizumab mafodotin-1MG
SDP
Catalog No. 503134766
Encompass_Preferred
Depatuxizumab Mafodotin (Depatux-M ABT-414) is an antibody-drug conjugate consisting of an EGFR IGg1 monoclonal antibody conjugated to the tubulin inhibitor monomethyl auristatin F via a stable maleimidocaproyl link Depatuxizumab Mafodotin is used in the treatment of recurrent EGFR-amplified Glioblastoma (GBM)
Catalog No. 50-313-4766 Supplier Selleck Chemical LLC Supplier No. D40421MG
Depatuxizumab Mafodotin (Depatux-M ABT-414) is an antibody-drug conjugate consisting of an EGFR IGg1 monoclonal antibody conjugated to the tubulin inhibitor monomethyl auristatin F via a stable maleimidocaproyl link Depatuxizumab Mafodotin is used in the treatment of recurrent EGFR-amplified Glioblastoma (GBM)