Learn More
Research Products International Corp DEPC, 200 ML
SDP
Supplier:  Research Products International Corp D43060200.0
Encompass_Preferred
Packaged as 2 x 100ml bottles.
Modification reagent for His and Tyr residues in proteins. Inhibitor of nucleases. DEPC treated water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by RNases.
Catalog No. 50-197-7078