Learn More
Research Products International Corp DEPC [Diethyl Pyrocarbonate], 25 Milliliters
SDP
Supplier:  Research Products International Corp D4306025.0
Encompass_Preferred
Modification reagent for His and Tyr residues in proteins. Inhibitor of nucleases. DEPC treated water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by RNases.
Catalog No. 50-213-289