Learn More
Biotang Inc DEPC TREATED WATER, 500 ML, WT-005,Sterile, Certified DNAse/RNAse Free Deionized water (18.2 megohm-cm+-.2) was treated with 0.01% DEPC and incubated at 37 degrees Celsius for 12 hr. Autoclaved.
Catalog No. 50152399 Shop All BIOTANG Inc Products
Encompass_Preferred
DEPC TREATED WATER, 500 ML, WT-005,Sterile, Certified DNAse/RNAse Free Deionized water (18.2 megohm-cm+-.2) was treated with 0.01% DEPC and incubated at 37 degrees Celsius for 12 hr. Autoclaved.
Catalog No. 50-152-399 Supplier Biotang Inc Supplier No. BTBB945
DEPC TREATED WATER, 500 ML, WT-005,Sterile, Certified DNAse/RNAse Free Deionized water (18.2 megohm-cm+-.2) was treated with 0.01% DEPC and incubated at 37 degrees Celsius for 12 hr. Autoclaved.