Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Water, DEPC Treated, 10 Liters
SDP

Catalog No. 50146832
Encompass_Preferred

DEPC Treated Water can be used to reconstitute nucleic acids and/or proteins. In addition, it can be used as a diluent or solvent for any molecular biology grade reagent.

DNase, RNase and Protease tested

DEPC Treated Water may be usefully employed in:
• RNA Applications (e.g., preparations of an RNA probe)
• DNA Applications (e.g., subcloning in pUC plasmids)
• Protein Applications (e.g., Western Blotting)

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered & Deionized
DNase, RNase, and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-146-832 Supplier Quality Biological Inc Supplier No. 351068151
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DEPC Treated Water can be used to reconstitute nucleic acids and/or proteins. In addition, it can be used as a diluent or solvent for any molecular biology grade reagent.

DNase, RNase and Protease tested

DEPC Treated Water may be usefully employed in:
• RNA Applications (e.g., preparations of an RNA probe)
• DNA Applications (e.g., subcloning in pUC plasmids)
• Protein Applications (e.g., Western Blotting)

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered & Deionized
DNase, RNase, and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.