Learn More
Kd Medical Inc DEPC ULTRA PURE WATER, 1L
SDP
Catalog No. 501031401
Encompass_Preferred
DEPC-treated (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by Rnases. Used in several Molecular Biology applications. One 1 Liter bottle.
Catalog No. 50-103-1401 Supplier Kd Medical Inc Supplier No. RGF3050
DEPC-treated (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by Rnases. Used in several Molecular Biology applications. One 1 Liter bottle.