Learn More
Kd Medical Inc DEPC ULTRA PURE WATER, 20L
SDP
Catalog No. 501031450
Encompass_Preferred
DEPC-treated (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by Rnases. Used in several Molecular Biology applications. One 20 liter cubitainer.
Catalog No. 50-103-1450 Supplier Kd Medical Inc Supplier No. RGJ0050
DEPC-treated (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by Rnases. Used in several Molecular Biology applications. One 20 liter cubitainer.