Learn More
Kd Medical Inc DEPC ULTRA PURE WATER, 4L
SDP
Catalog No. 501031435
Encompass_Preferred
DEPC-treated (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by Rnases. Used in several Molecular Biology applications. One 4 Liter cubitainer.
Catalog No. 50-103-1435 Supplier Kd Medical Inc Supplier No. RGH0050
DEPC-treated (and therefore RNase-free) water is used in handling of RNA in the laboratory to reduce the risk of RNA being degraded by Rnases. Used in several Molecular Biology applications. One 4 Liter cubitainer.