Learn More
Aviva Systems Biology Corp DEPDC1 Antibody - middle region (ARP56317_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56317_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DEPDC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human DEPDC1. Alternate names: DEP.8, DEPDC1-V2, DEPDC1A, FLJ20354, SDP35ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-139