Learn More
Aviva Systems Biology Corp DEPDC7 Antibody - N-terminal region (ARP51708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DEPDC7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DEPDC7. Alternate names: dJ85M6.4, TR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-586