Learn More
Crescent Chemical Co Inc DEPEX
SDP
Supplier:  Crescent Chemical Co Inc SE18243.02
Encompass_Preferred
DePeX Mounting medium for histology. Refractive index 1.52 - 1.53. Neutral solution of polystyrene and plasticizers in xylene. Size - 500ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 18243.02
Catalog No. 50-247-471