Learn More
Crescent Chemical Co Inc DEPEX
SDP
Catalog No. 50247470
Encompass_Preferred
DePeX Mounting medium for histology. Refractive index 1.52 - 1.53. Neutral solution of polystyrene and plasticizers in xylene. Size - 100ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 18243.01
Catalog No. 50-247-470 Supplier Crescent Chemical Co Inc Supplier No. SE18243.01
DePeX Mounting medium for histology. Refractive index 1.52 - 1.53. Neutral solution of polystyrene and plasticizers in xylene. Size - 100ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 18243.01