Learn More
Electron Microscopy Sciences Depex Mounting Medium 100 ML
SDP
Catalog No. 50980371
Encompass_Preferred
A colorless, neutral, xylene miscible mountant which is suitable for preparations where Canada Balsam is usually used.
Catalog No. 50-980-371 Supplier Electron Microscopy Sciences Supplier No. 13514
A colorless, neutral, xylene miscible mountant which is suitable for preparations where Canada Balsam is usually used.