Learn More
Electron Microscopy Sciences DEPEX Mounting Media, 500ml
SDP
Catalog No. 50980372
Encompass_Preferred
DEPEX Mounting Media is a colorless, neutral, xylene miscible mountant which is suitable for preparations where Canada Balsam is usually used. It comes in the amount of 500ml.
Catalog No. 50-980-372 Supplier Electron Microscopy Sciences Supplier No. 13515
DEPEX Mounting Media is a colorless, neutral, xylene miscible mountant which is suitable for preparations where Canada Balsam is usually used. It comes in the amount of 500ml.