Learn More
Enzo Life Sciences DEPMPO (500mg). CAS: 157230-67-6
SDP
Supplier:  Enzo Life Sciences ALX430093M500
Encompass_Preferred
Most efficient spin trap for the in vitro and in vivo detection of O-, N-, S- and C-centered free radicals. Has a longer life-time than DMPO (ALX-430-090). Can distinguish between superoxide-dependent and independent mechanisms that lead to the hydroxyl radical. Less lipophilic (Kp=0.16) than DIPPMPO (ALX-430-119). Alternative name: 5-(Diethoxyphosphoryl)-5-methyl-1-pyrroline-N-oxide, 2-Diethylphosphono-2-methyl-3,4-dihydro-2H-pyrrole 1-oxide, (2-Methyl-3,4-dihydro-1-oxide-2H-pyrrol-2-yl)diethylphosphonate. Purity: ≥99% (TLC). Appearance: Colorless to light yellow oil. Solubility: Soluble in water, methylene chloride or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9196