Learn More
Cayman Chemical DEPMPO-bIotIn 250ug
SDP
Catalog No. 502954771
Encompass_Preferred
A biotinylated form of DEPMPO, which is used to spin trap reactive O-, N-, S-, and C-centered radicals; offers monitored biodistribution in cells, tissues and organs when used with an avidin-conjugated reporter; binds free radicals on proteins, producing adducts that can be analyzed via the biotin tag.
Catalog No. 50-295-4771 Supplier Cayman Chemical Supplier No. 13251250 UG
A biotinylated form of DEPMPO, which is used to spin trap reactive O-, N-, S-, and C-centered radicals; offers monitored biodistribution in cells, tissues and organs when used with an avidin-conjugated reporter; binds free radicals on proteins, producing adducts that can be analyzed via the biotin tag.