Learn More
Abcam DEPMPO, Spin trap, 500MG
Catalog No. 502705959
Encompass_Preferred
MW 235.22 Da, Purity >99%. Spin trap. Longer half-life derivative of DMPO (ab144610). Distinguishes between superoxide-dependent and independent mechanisms that lead to the hydroxyl radical Detects O-, N-, S- and C-centered free radicals in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5959 Supplier Abcam Supplier No. AB144240500MG
MW 235.22 Da, Purity >99%. Spin trap. Longer half-life derivative of DMPO (ab144610). Distinguishes between superoxide-dependent and independent mechanisms that lead to the hydroxyl radical Detects O-, N-, S- and C-centered free radicals in vivo. .
The product is subject to the following: Abcam Restricted Use Statement