Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Depressine | 176182-06-2 | 1 MG
SDP

Catalog No. 5000419592
Encompass_Preferred

Depressine is an iridoid glycoside isolated from the methanol extract of the aerial parts of Gentiana depressa. It can be used to reduce oxidative DNA base damage and strand breaks that are prone to occur during the preparation of silver nanoparticles (AgNPs).

  • Iridoid glycoside
  • Isolated from Gentiana depressa
  • Reduces oxidative DNA base damage
  • Reduces strand breaks during silver nanoparticle preparation
  • Molecular weight: 688.63
  • Molecular formula: C30H40O18
  • Appearance: solid, white to light yellow
  • Purity: 99.84%

Catalog No. 50-004-19592 Supplier Medchemexpress LLC Supplier No. HYN50701MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Depressine is an iridoid glycoside isolated from the methanol extract of the aerial parts of Gentiana depressa. It can be used to reduce oxidative DNA base damage and strand breaks that are prone to occur during the preparation of silver nanoparticles (AgNPs).

  • Iridoid glycoside
  • Isolated from Gentiana depressa
  • Reduces oxidative DNA base damage
  • Reduces strand breaks during silver nanoparticle preparation
  • Molecular weight: 688.63
  • Molecular formula: C30H40O18
  • Appearance: solid, white to light yellow
  • Purity: 99.84%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.