Learn More
Medchemexpress LLC HY-U00190 5mg Medchemexpress, Deprodone propionate CAS:20424-00-4 Purity:>98%
SDP
Catalog No. 501970627
Encompass_Preferred
Medchemexpress, HY-U00190 5mg Deprodone propionate CAS:20424-00-4 Deprodone propionate (RD20000) is a corticosteroid which is obtained by esterifying with propionic acid the 17-position of the prednisolone skeleton and deoxidating its 21-position. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0627 Supplier Medchemexpress LLC Supplier No. HYU001905MG
Medchemexpress, HY-U00190 5mg Deprodone propionate CAS:20424-00-4 Deprodone propionate (RD20000) is a corticosteroid which is obtained by esterifying with propionic acid the 17-position of the prednisolone skeleton and deoxidating its 21-position. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.