Learn More
BURKLE INC Depth catch net, PP/PA, LxW 7.48x16.93 in. (190x430 mm), TeleScoop
Catalog No. 5000025668
Encompass_Preferred
Catch net for taking solid and organic samples from water courses.
Catalog No. 50-000-25668 Supplier BURKLE INC Supplier No. 53545010
Catch net for taking solid and organic samples from water courses.