Learn More
Med Vet International Depth Gauge, (2.7-4.0mm) Range 6-40mm, J0410
Catalog No. 5000582309
Encompass_Preferred
Accurate measurement of hole depth is essential to select the appropriate screw size. A range of gauges is available for the various screw sizes. Hole depth is read from the instrument. Remember to add plate thickness if necessary.
Catalog No. 50-005-82309 Supplier Med Vet International Supplier No. JOR410
Accurate measurement of hole depth is essential to select the appropriate screw size. A range of gauges is available for the various screw sizes. Hole depth is read from the instrument. Remember to add plate thickness if necessary.