Learn More
Bioworld Depurination Solution, 2 L
Catalog No. 50196416
Encompass_Preferred
- for Biotechnology Grade-Used for depurination of DNA in DNA blots- Recipe : Contains 0.25M HCIpH @25C - <=2.0 Solution-Clear & Colorless (P/F)- Pass
Catalog No. 50-196-416 Supplier Bioworld Supplier No. 102500002
- for Biotechnology Grade-Used for depurination of DNA in DNA blots- Recipe : Contains 0.25M HCIpH @25C - <=2.0 Solution-Clear & Colorless (P/F)- Pass