Learn More
Bioworld Depurination Solution, 1 L
Catalog No. 50196415
Encompass_Preferred
- for Biotechnology Grade-Used for depurination of DNA in DNA blots- Recipe : Contains 0.25M HCIpH @25C - <=2.0 Solution-Clear & Colorless (P/F)- Pass
Catalog No. 50-196-415 Supplier Bioworld Supplier No. 102500001
- for Biotechnology Grade-Used for depurination of DNA in DNA blots- Recipe : Contains 0.25M HCIpH @25C - <=2.0 Solution-Clear & Colorless (P/F)- Pass