Learn More
Grainger Deputy Deluxe Shirt Brown 18 in
Catalog No. 502751214
Encompass_Preferred
The Grainger 14K955 is a Westward brand 1/4-inch drive click torque wrench that measures torque in inch pounds. It has a range of 10 to 150 inch pounds and a +/- 4% accuracy. The wrench has a reversible ratchet head, knurled grip, and a positive lock with torque adjustment. It is made of chrome vanadium steel and comes with a plastic storage case. The wrench is 20 inches long and weighs 1.75 pounds. It can be calibrated and is covered by a lifetime warranty.
Catalog No. 50-275-1214 Supplier Grainger Supplier No. 14K955
The Grainger 14K955 is a Westward brand 1/4-inch drive click torque wrench that measures torque in inch pounds. It has a range of 10 to 150 inch pounds and a +/- 4% accuracy. The wrench has a reversible ratchet head, knurled grip, and a positive lock with torque adjustment. It is made of chrome vanadium steel and comes with a plastic storage case. The wrench is 20 inches long and weighs 1.75 pounds. It can be calibrated and is covered by a lifetime warranty.