Learn More
TARGETMOL CHEMICALS INC Dequalinium chloride 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0979200MG
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dequalinium chloride (Decabis) , a topical bacteriostat, is a selective blocker of apamin-sensitive K+ channels. It is used in wound dressings and mouth infections and may also have antifungal action, but may cause skin ulceration. Purity 99.58%
Catalog No. 50-288-0528