Learn More
Medchemexpress LLC Dequalinium chloride | 522-51-0 | 99.7% | 527.57 g/mol | C30H40Cl2N4 | 10 MG
SDP
Catalog No. 5000299032
Encompass_Preferred
Dequalinium chloride is a cationic, lipophilic antiseptic and research reagent that functions as an apamin-sensitive potassium channel blocker and an α7 nicotinic acetylcholine receptor antagonist. It is used in biochemical and pharmacological studies and may be applied in both in vitro and in vivo experiments.
Catalog No. 50-002-99032 Supplier Medchemexpress LLC Supplier No. HYB056710MG
Dequalinium chloride is a cationic, lipophilic antiseptic and research reagent that functions as an apamin-sensitive potassium channel blocker and an α7 nicotinic acetylcholine receptor antagonist. It is used in biochemical and pharmacological studies and may be applied in both in vitro and in vivo experiments.