Learn More
Electron Microscopy Sciences DER 332/732 Kit
SDP
Catalog No. 50980382
Encompass_Preferred
Three different mixtures: one for relatively soft blocks, another for harder blocks for collagenous tissue, and a tougher one if required. Reference: Lockwood, W.R. Anatomical Record, 150, 129 (1964).
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA. BDMA is less viscous, has a longer shelf life, and offers better penetration. When using BDMA in place of DMP-30 you must vary the amount: DMP-30: 1.5-2%; BDMA: 2.5-3%.
Catalog No. 50-980-382 Supplier Electron Microscopy Sciences Supplier No. 14000
Three different mixtures: one for relatively soft blocks, another for harder blocks for collagenous tissue, and a tougher one if required. Reference: Lockwood, W.R. Anatomical Record, 150, 129 (1964).
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA. BDMA is less viscous, has a longer shelf life, and offers better penetration. When using BDMA in place of DMP-30 you must vary the amount: DMP-30: 1.5-2%; BDMA: 2.5-3%.