Learn More
Electron Microscopy Sciences DER 732 Resin 225 ML
SDP
Supplier:  Electron Microscopy Sciences 13010
Encompass_Preferred
Used as a flexibilizer in Maraglas 655-epoxy formulation. Relatively low Viscosity (55-100 at 25 degree C). (Erlandson, J. Cell Bio., 22,704 1964).
Catalog No. 50-980-344