Learn More
Electron Microscopy Sciences DER 736 Resin 225 ML
SDP
Catalog No. 50980342
Encompass_Preferred
A light colored, flexible epoxy. Used to simplify infiltration in combination with Epon 812 (Embed 812). Kushida, J. Electron Micro., 16,278 (1964)
Catalog No. 50-980-342 Supplier Electron Microscopy Sciences Supplier No. 13000
A light colored, flexible epoxy. Used to simplify infiltration in combination with Epon 812 (Embed 812). Kushida, J. Electron Micro., 16,278 (1964)