Learn More
Bioworld DER- F1 PROTEIN (RECOMBINANT)
Catalog No. 50253148
Encompass_Preferred
Recombinant Dermatophagoides F1 Mosaic Protein (Der-F1) Source: E.Coli Background: DerF1 is a thiol protease that hydrolyzes proteins, with a preference for Phe or basic residues. DERF1 is a C1 peptidase family member. DERF1 has exte...
Catalog No. 50-253-148 Supplier Bioworld Supplier No. 220600511
Recombinant Dermatophagoides F1 Mosaic Protein (Der-F1) Source: E.Coli Background: DerF1 is a thiol protease that hydrolyzes proteins, with a preference for Phe or basic residues. DERF1 is a C1 peptidase family member. DERF1 has exte...