Learn More
Medchemexpress LLC Der p 23 protein | >95.0% | 10 UG
SDP
Catalog No. 5000291527
Encompass_Preferred
Recombinant Der p 23 protein (residues A22-T90) is a C-terminal His-tagged, HEK293-expressed allergen supplied as a lyophilized powder for immunological assays, antibody generation, and allergy research.
  • Recombinant major mite allergen suitable for allergy research.
  • Expressed in HEK293 for eukaryotic post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • High purity (>95.0%) confirmed by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for sensitive immunoassays.
  • Lyophilized format for long-term storage and convenient handling.
  • Includes storage and reconstitution guidance to preserve activity.
Catalog No. 50-002-91527 Supplier Medchemexpress LLC Supplier No. HYP7528910UG
Recombinant Der p 23 protein (residues A22-T90) is a C-terminal His-tagged, HEK293-expressed allergen supplied as a lyophilized powder for immunological assays, antibody generation, and allergy research.
  • Recombinant major mite allergen suitable for allergy research.
  • Expressed in HEK293 for eukaryotic post-translational modifications.
  • C-terminal His tag enables affinity purification and detection.
  • High purity (>95.0%) confirmed by reducing SDS-PAGE.
  • Low endotoxin (<1 EU/μg) suitable for sensitive immunoassays.
  • Lyophilized format for long-term storage and convenient handling.
  • Includes storage and reconstitution guidance to preserve activity.