Learn More
Medchemexpress LLC Der p 23 protein, Dermatophagoides pteronyssinus | >95.0% | 100 UG
SDP
Catalog No. 5000291525
Encompass_Preferred
Der p 23 is a recombinant major allergen from the house dust mite Dermatophagoides pteronyssinus, expressed in HEK293 cells and supplied as a lyophilized C-terminal His-tagged protein. It is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 19 kDa, and endotoxin <1 EU/μg. The product is formulated in PBS (pH 7.4) and is recommended to be stored at -20°C; add carrier protein for long-term storage.
  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (>95%) suitable for immunological assays.
  • Low endotoxin level (<1 EU/μg) for sensitive applications.
  • Lyophilized formulation; PBS-based, recommended -20°C storage.
Catalog No. 50-002-91525 Supplier Medchemexpress LLC Supplier No. HYP75289100UG
Der p 23 is a recombinant major allergen from the house dust mite Dermatophagoides pteronyssinus, expressed in HEK293 cells and supplied as a lyophilized C-terminal His-tagged protein. It is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 19 kDa, and endotoxin <1 EU/μg. The product is formulated in PBS (pH 7.4) and is recommended to be stored at -20°C; add carrier protein for long-term storage.
  • Recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for purification and detection.
  • High purity (>95%) suitable for immunological assays.
  • Low endotoxin level (<1 EU/μg) for sensitive applications.
  • Lyophilized formulation; PBS-based, recommended -20°C storage.