Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Der p 23 protein (Dermatophagoides pteronyssinus) | >95.0% | 50 UG
SDP

Catalog No. 5000292598
Encompass_Preferred

Recombinant Der p 23 protein derived from the house dust mite Dermatophagoides pteronyssinus. Supplied lyophilized in a 50 μg vial, this sequence-verified allergen is intended for research use in allergy and immunology studies where defined recombinant antigen is required.

  • Recombinant allergen suitable for allergy and immunology research.
  • Expressed in HEK293 cells with a C-terminal His tag.
  • Approximately 19 kDa; sequence verified.
  • Lyophilized from PBS (pH 7.4) for stability and convenience.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Store lyophilized at -20°C; freeze aliquots for long-term storage.
  • Recommended minimum reconstitution concentration 100 μg/mL in ddH2O.

Catalog No. 50-002-92598 Supplier Medchemexpress LLC Supplier No. HYP7528950UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Der p 23 protein derived from the house dust mite Dermatophagoides pteronyssinus. Supplied lyophilized in a 50 μg vial, this sequence-verified allergen is intended for research use in allergy and immunology studies where defined recombinant antigen is required.

  • Recombinant allergen suitable for allergy and immunology research.
  • Expressed in HEK293 cells with a C-terminal His tag.
  • Approximately 19 kDa; sequence verified.
  • Lyophilized from PBS (pH 7.4) for stability and convenience.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Store lyophilized at -20°C; freeze aliquots for long-term storage.
  • Recommended minimum reconstitution concentration 100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.