Learn More
Bioworld DER-P1 PROTEIN (RECOMBINANT)
Supplier:  Bioworld 220600521
Encompass_Preferred
Recombinant Dermatophagoides P1 Protein (Der-p1) Source: E.Coli Background: DERP1 is a thiol protease, with a preference for substrates with a large hydrophobic side chain in the P2 position, or with basic residues. DERP1 is a C1 pep...
Catalog No. 50-253-149