Learn More
Medchemexpress LLC Deramciclane | 120444-71-5 | MFCD00865305 | 98.0% | 301.47 g/mol | C20H31NO | 50 MG
SDP
Catalog No. 5000248507
Encompass_Preferred
Deramciclane is a research compound (CAS 120444-71-5) that functions as an antagonist/inverse agonist at serotonin 5-HT2A and 5-HT2C receptors. It has formula C20H31NO and a molecular weight of 301.47 g/mol, and is supplied for laboratory research in small pack sizes (for example, 50 MG). Reported purity is approximately 98%.
  • Acts as 5-HT2A and 5-HT2C antagonist/inverse agonist.
  • Applicable for pharmacology and neurobiology research into serotonergic pathways.
  • Molecular formula C20H31NO and molecular weight 301.47 g/mol.
  • Typical purity around 98%, with batch-specific certificates available.
  • Packaged in small quantities suitable for bench-scale experiments.
  • Accompanied by a safety data sheet and batch information for traceability.
Catalog No. 50-002-48507 Supplier Medchemexpress LLC Supplier No. HY10163050MG
Deramciclane is a research compound (CAS 120444-71-5) that functions as an antagonist/inverse agonist at serotonin 5-HT2A and 5-HT2C receptors. It has formula C20H31NO and a molecular weight of 301.47 g/mol, and is supplied for laboratory research in small pack sizes (for example, 50 MG). Reported purity is approximately 98%.
  • Acts as 5-HT2A and 5-HT2C antagonist/inverse agonist.
  • Applicable for pharmacology and neurobiology research into serotonergic pathways.
  • Molecular formula C20H31NO and molecular weight 301.47 g/mol.
  • Typical purity around 98%, with batch-specific certificates available.
  • Packaged in small quantities suitable for bench-scale experiments.
  • Accompanied by a safety data sheet and batch information for traceability.