Learn More
Medchemexpress LLC Deramciclane | 120444-71-5 | MFCD00865305 | 98.1% | 301.47 g/mol | C20H31NO | 100 MG
SDP
Catalog No. 5000248455
Encompass_Preferred
Deramciclane is a research-grade small molecule that acts at serotonin 5-HT2A and 5-HT2C receptors, reported to function as an antagonist and inverse agonist. It is used in pharmacology and preclinical studies to investigate anxiolytic mechanisms, receptor binding, and signaling pathways. Key chemical identifiers include CAS 120444-71-5, chemical formula C20H31NO, and a molecular weight of about 301.47 g/mol.
  • High purity suitable for research applications.
  • Characterized by standard analytical methods.
  • Suitable for in vitro receptor binding and functional assays.
  • Supplied as a weighed solid for accurate dosing.
  • Stable when stored under recommended laboratory conditions.
Catalog No. 50-002-48455 Supplier Medchemexpress LLC Supplier No. HY101630100MG
Deramciclane is a research-grade small molecule that acts at serotonin 5-HT2A and 5-HT2C receptors, reported to function as an antagonist and inverse agonist. It is used in pharmacology and preclinical studies to investigate anxiolytic mechanisms, receptor binding, and signaling pathways. Key chemical identifiers include CAS 120444-71-5, chemical formula C20H31NO, and a molecular weight of about 301.47 g/mol.
  • High purity suitable for research applications.
  • Characterized by standard analytical methods.
  • Suitable for in vitro receptor binding and functional assays.
  • Supplied as a weighed solid for accurate dosing.
  • Stable when stored under recommended laboratory conditions.