Learn More
Medchemexpress LLC Deramciclane | 120444-71-5 | MFCD00865305 | 98.1% | 301.47 g·mol⁻¹ | C20H31NO | 25 MG
SDP
Catalog No. 5000245684
Encompass_Preferred
Deramciclane is a serotonin (5-HT) receptor modulator used in pharmacological research, particularly in studies of anxiolytic activity. It is supplied as a high-purity research reagent in small laboratory pack sizes for in vitro and in vivo experiments.
  • High purity (≈98.1%) appropriate for in vitro and in vivo studies.
  • Chemical formula C20H31NO and molecular weight 301.47 g·mol⁻¹.
  • Suitable for studies of 5-HT receptor pharmacology and behavioral assays.
  • Available in small pack sizes commonly used in research, for example 25 mg.
Catalog No. 50-002-45684 Supplier Medchemexpress LLC Supplier No. HY10163025MG
Deramciclane is a serotonin (5-HT) receptor modulator used in pharmacological research, particularly in studies of anxiolytic activity. It is supplied as a high-purity research reagent in small laboratory pack sizes for in vitro and in vivo experiments.
  • High purity (≈98.1%) appropriate for in vitro and in vivo studies.
  • Chemical formula C20H31NO and molecular weight 301.47 g·mol⁻¹.
  • Suitable for studies of 5-HT receptor pharmacology and behavioral assays.
  • Available in small pack sizes commonly used in research, for example 25 mg.