Learn More
TARGETMOL CHEMICALS INC DERAZANTINIB 5MG
SDP
Catalog No. 502578811
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Derazantinib (ARQ-087) (ARQ-087) is an ATP competitive tyrosine kinase inhibitor. It exhibits potent activity against FGFR1/FGFR2/FGFR3 chondrocytes (IC50s 4.5 nM/1.8 nM/4.5 nM). purity: 99%
Catalog No. 50-257-8811 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ02285MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Derazantinib (ARQ-087) (ARQ-087) is an ATP competitive tyrosine kinase inhibitor. It exhibits potent activity against FGFR1/FGFR2/FGFR3 chondrocytes (IC50s 4.5 nM/1.8 nM/4.5 nM). purity: 99%