Learn More
Selleck Chemical LLC Derazantinib(ARQ-087) 2mg 1234356-69-4
SDP
Catalog No. 501948284
Encompass_Preferred
Derazantinib(ARQ-087) is an orally bioavailable inhibitor of the fibroblast growth factor receptor (FGFR) with IC50 values of 1.8 nM for FGFR2, and 4.5 nM for FGFR1 and 3, showing lower potency for FGFR4 (IC50=34 nM). It also inhibits RET, DDR2, PDGFR?, VEGFR and KIT. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8284 Supplier Selleck Chemical LLC Supplier No. S86092MG
Derazantinib(ARQ-087) is an orally bioavailable inhibitor of the fibroblast growth factor receptor (FGFR) with IC50 values of 1.8 nM for FGFR2, and 4.5 nM for FGFR1 and 3, showing lower potency for FGFR4 (IC50=34 nM). It also inhibits RET, DDR2, PDGFR?, VEGFR and KIT. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.