Learn More
Medchemexpress LLC HY-19981A 2mg Medchemexpress, Derazantinib Racemate CAS: Purity:>98%
SDP
Catalog No. 501932847
Encompass_Preferred
Medchemexpress, HY-19981A 2mg Derazantinib Racemate CAS: Derazantinib Racemate is the racemate of derazantinib; Derazantinib is a potent inhibitor of FGFR1-3 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2847 Supplier Medchemexpress LLC Supplier No. HY19981A2MG
Medchemexpress, HY-19981A 2mg Derazantinib Racemate CAS: Derazantinib Racemate is the racemate of derazantinib; Derazantinib is a potent inhibitor of FGFR1-3 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.