Learn More
Cayman Chemical Derenofylline, Purity: ≥98%, CAS: 251945-92-3, 10 mg
SDP
Catalog No. 502940371
Encompass_Preferred
An adenosine A1 receptor antagonist (Ki = 1 nM); selective for adenosine A1 over A2A, A2B, and A3 receptors (Kis = 398, 3,981, and 200 nM, respectively); decreases adenosine A1 receptor-mediated adenosine-induced bradycardia in rats (ED50 = 0.49 mg/kg) but reduces A2 receptor-mediated adenosine-induced hypotension by only 44.6% at an intravenous dose of 5 mg/kg; prevents increases in heart levels of collagen I and III in nephrectomized rats at 10 mg/kg per day; reduces relative plaque counts in a Zika virus plaque-forming assay in A549 cells (IC50 = 58.6 nM) in an adenosine A1 receptor-independent manner
Catalog No. 50-294-0371 Supplier Cayman Chemical Supplier No. 2766610 MG
An adenosine A1 receptor antagonist (Ki = 1 nM); selective for adenosine A1 over A2A, A2B, and A3 receptors (Kis = 398, 3,981, and 200 nM, respectively); decreases adenosine A1 receptor-mediated adenosine-induced bradycardia in rats (ED50 = 0.49 mg/kg) but reduces A2 receptor-mediated adenosine-induced hypotension by only 44.6% at an intravenous dose of 5 mg/kg; prevents increases in heart levels of collagen I and III in nephrectomized rats at 10 mg/kg per day; reduces relative plaque counts in a Zika virus plaque-forming assay in A549 cells (IC50 = 58.6 nM) in an adenosine A1 receptor-independent manner