Learn More
Medchemexpress LLC Derenofylline | 251945-92-3 | 99.9% | 308.38 | 50 MG
SDP
Catalog No. 5000360127
Encompass_Preferred
Derenofylline (SLV 320) is a potent, selective, and orally active adenosine A1 receptor antagonist. It exhibits Ki values of 1 nM for human A1, 200 nM for A3, and 398 nM for A2A receptors. This compound suppresses cardiac fibrosis and attenuates albuminuria without affecting blood pressure in rats. It has also shown to inhibit TGF-β1-induced myofibroblast transformation in vitro and reduce myocardial fibrosis in vivo.
  • Potent, selective, and orally active adenosine A1 receptor antagonist
  • Suppresses cardiac fibrosis
  • Attenuates albuminuria
  • Inhibits TGF-β1-induced myofibroblast transformation (in vitro)
  • Reduces myocardial fibrosis in 5/6 nephrectomy rats (in vivo)
Catalog No. 50-003-60127 Supplier Medchemexpress LLC Supplier No. HY1485850MG
Derenofylline (SLV 320) is a potent, selective, and orally active adenosine A1 receptor antagonist. It exhibits Ki values of 1 nM for human A1, 200 nM for A3, and 398 nM for A2A receptors. This compound suppresses cardiac fibrosis and attenuates albuminuria without affecting blood pressure in rats. It has also shown to inhibit TGF-β1-induced myofibroblast transformation in vitro and reduce myocardial fibrosis in vivo.
  • Potent, selective, and orally active adenosine A1 receptor antagonist
  • Suppresses cardiac fibrosis
  • Attenuates albuminuria
  • Inhibits TGF-β1-induced myofibroblast transformation (in vitro)
  • Reduces myocardial fibrosis in 5/6 nephrectomy rats (in vivo)